siRNA Details
Gene Symbol: Msh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031058.1 | NP_112320.1 | 10 | 1566 | ||
GenBank mRNA | BC161846.1 | - | 10 | 1582 | 9029023 | |
X93591.1 | - | 10 | 1566 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Msh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031058.1 | NP_112320.1 | 10 | 1566 | ||
GenBank mRNA | BC161846.1 | - | 10 | 1582 | 9029023 | |
X93591.1 | - | 10 | 1566 |