siRNA Details
Gene Symbol: Cdo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052809.1 | NP_434696.1 | 4 | 610 | ||
GenBank mRNA | BC070509.1 | - | 4 | 601 | 7100321 | |
FQ218376.1 | - | 3 | 247 | |||
FQ219282.1 | - | 3 | 247 | |||
M35266.1 | - | 4 | 610 |

More Information
Other Species Targeted

Gene Ontology Categories:
