siRNA Details
Gene Symbol: Rpl10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031065.1 | NP_112327.1 | 4 | 321 | ||
GenBank mRNA | BC058468.1 | - | 4 | 328 | 6919792 | |
FQ221330.1 | - | 4 | 321 | |||
FQ221725.1 | - | 4 | 324 | |||
FQ222317.1 | - | 4 | 326 | |||
X93352.1 | - | 4 | 321 |
Gene Symbol: LOC100910579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246873.1 | XP_006246935.1 | 2 | 454 |
Gene Symbol: LOC103690888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773474.1 | XP_008771696.1 | 1 | 340 |
Gene Symbol: RGD1566137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244660.1 | XP_006244722.1 | 1 | 328 | ||
XM_217361.6 | XP_217361.4 | 1 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
