siRNA Details
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031081.1 | NP_112343.1 | 13 | 1492 | ||
XM_006246036.2 | XP_006246098.1 | 13 | 3344 | |||
GenBank mRNA | Y15748.1 | - | 13 | 1492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
