siRNA Details
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031081.1 | NP_112343.1 | 12, 13 | 1476 | ||
XM_006246036.2 | XP_006246098.1 | 12, 13 | 3328 | |||
GenBank mRNA | Y15748.1 | - | 12, 13 | 1476 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001286662, NM_011062, NM_001080773, AY062008, AF079535, and others.
Gene Ontology Categories:
