siRNA Details
Gene Symbol: Psen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031087.2 | NP_112349.2 | 7 | 951 | ||
XM_006250405.2 | XP_006250467.1 | 9 | 1102 | |||
XM_006250407.2 | XP_006250469.1 | 8 | 1012 | |||
XM_006250408.2 | XP_006250470.1 | 7 | 927 | |||
XM_006250410.2 | XP_006250472.1 | 8 | 1695 | |||
XM_008769876.1 | XP_008768098.1 | 8 | 1694 | |||
XM_008769877.1 | XP_008768099.1 | 9 | 1242 | |||
XM_008769878.1 | XP_008768100.1 | 7 | 2320 | |||
GenBank mRNA | AB004454.1 | - | 6 | 861 | ||
BC078805.1 | - | 7 | 951 | 7109917 | ||
D83700.1 | - | 6 | 854 | |||
FQ219614.1 | - | 9 | 1072 | |||
X99267.1 | - | 7 | 974 |
