siRNA Details
Gene Symbol: Rpl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031103.1 | NP_112365.1 | 4 | 356 | ||
GenBank mRNA | BC058135.1 | - | 4 | 356 | 6918377 | |
FQ209882.1 | - | 2 | 185 | |||
FQ211447.1 | - | 2 | 185 | |||
FQ211816.1 | - | 2 | 185 | |||
FQ221779.1 | - | 2 | 185 | |||
FQ221820.1 | - | 2 | 185 | |||
FQ222282.1 | - | 4 | 356 | |||
J02650.1 | - | 4 | 356 |
Gene Symbol: RGD1560099 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_228526.7 | XP_228526.4 | 1 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
