siRNA Details
Gene Symbol: Rps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031108.3 | NP_112370.2 | 5 | 553 | ||
GenBank mRNA | BC060560.1 | - | 5 | 558 | 6922405 | |
FQ229992.1 | - | 5 | 513 | |||
X66370.1 | - | 4 | 487 |
Gene Symbol: LOC103689992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758836.1 | XP_008757058.1 | 583 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
