siRNA Details
Gene Symbol: Rps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031109.1 | NP_112371.1 | 3 | 236 | ||
GenBank mRNA | BC058141.1 | - | 3 | 224 | 6921499 | |
FQ210135.1 | - | 3 | 226 | |||
FQ210550.1 | - | 3 | 227 | |||
FQ217175.1 | - | 3 | 225 | |||
FQ217183.1 | - | 3 | 226 | |||
FQ221004.1 | - | 3 | 226 | |||
FQ221394.1 | - | 3 | 226 | |||
FQ221397.1 | - | 3 | 226 | |||
FQ221701.1 | - | 3 | 226 | |||
FQ221937.1 | - | 3 | 226 | |||
FQ222456.1 | - | 3 | 231 | |||
FQ222595.1 | - | 3 | 229 | |||
FQ222615.1 | - | 3 | 228 | |||
FQ222830.1 | - | 3 | 226 | |||
FQ223309.1 | - | 3 | 228 | |||
FQ223371.1 | - | 3 | 217 | |||
FQ224021.1 | - | 3 | 226 | |||
FQ228540.1 | - | 3 | 226 | |||
X13549.1 | - | 3 | 236 |
Gene Symbol: Rpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ229603.1 | - | 226 |
Gene Symbol: Rps10l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729623.2 | XP_002729669.1 | 1 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
