siRNA Details
Gene Symbol: Stx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031125.1 | NP_112387.1 | 8, 9 | 742 | ||
XM_006230345.2 | XP_006230407.1 | 8, 9 | 1249 | |||
XM_006230346.2 | XP_006230408.1 | 8, 9 | 1274 | |||
GenBank mRNA | L20821.1 | - | 8, 9 | 742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
