siRNA Details
Gene Symbol: Serpina7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257307.2 | XP_006257369.1 | 3 | 681 | ||
XM_006257308.2 | XP_006257370.1 | 2 | 1938 | |||
XM_008773465.1 | XP_008771687.1 | 2 | 1936 | |||
XM_008773466.1 | XP_008771688.1 | 3 | 674 | |||
XM_343823.6 | XP_343824.4 | 2 | 610 | |||
GenBank mRNA | M63991.1 | - | 1 | 502 |
