siRNA Details
Gene Symbol: Tceb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031129.3 | NP_112391.1 | 3 | 192 | ||
XM_008767596.1 | XP_008765818.1 | 2 | 198 | |||
GenBank mRNA | BC058463.1 | - | 3 | 196 | 6921630 | |
FQ212271.1 | - | 2 | 168 | |||
L42855.1 | - | 2 | 152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
