siRNA Details
Gene Symbol: Klf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031135.2 | NP_112397.1 | 3 | 614 | ||
GenBank mRNA | BC097309.1 | - | 3 | 614 | 7387595 | |
U78875.1 | - | 3 | 883 | |||
U88630.1 | - | 2 | 523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
