siRNA Details
Gene Symbol: Ube2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031138.2 | NP_112400.1 | 3, 4 | 260 | ||
XM_006246343.2 | XP_006246405.1 | 4, 5 | 285 | |||
GenBank mRNA | BC070946.1 | - | 3, 4 | 253 | 7103580 | |
DN931313.1 | - | 3, 4 | 290 | |||
FQ216310.1 | - | 1, 2 | 40 | |||
FQ222428.1 | - | 3, 4 | 277 | |||
FQ229894.1 | - | 3, 4 | 271 | |||
M62388.1 | - | 3, 4 | 202 |
Gene Symbol: LOC103694902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773893.1 | XP_008772115.1 | 4, 5 | 284 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009458, AK152645, AK147785, AK169229, AK010432, and others.
Gene Ontology Categories:
