siRNA Details
Gene Symbol: Cib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031145.1 | NP_112407.1 | 3 | 195 | ||
GenBank mRNA | AF136585.1 | - | 3 | 195 | ||
BC091143.1 | - | 3 | 181 | 7308828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
