siRNA Details
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031151.2 | NP_112413.2 | 2, 3 | 332 | ||
GenBank mRNA | BC063165.1 | - | 2, 3 | 332 | 6918633 | |
DQ402951.1 | - | 1, 2 | 166 | |||
FQ213494.1 | - | 2, 3 | 285 | |||
FQ213817.1 | - | 2, 3 | 249 | |||
FQ215385.1 | - | 2, 3 | 292 | |||
FQ215647.1 | - | 2, 3 | 293 | |||
FQ216782.1 | - | 2, 3 | 323 | |||
FQ217232.1 | - | 2, 3 | 284 | |||
FQ224847.1 | - | 2, 3 | 229 | |||
FQ224979.1 | - | 2, 3 | 311 | |||
X04240.1 | - | 2, 3 | 273 |

More Information
Other Species Targeted

Gene Ontology Categories:
