siRNA Details
Gene Symbol: Sharpin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031153.2 | NP_112415.1 | 2 | 590 | ||
GenBank mRNA | AF203906.1 | - | 2 | 264 | ||
BC083553.1 | - | 2 | 590 | 7189111 | ||
FQ213170.1 | - | 2 | 208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
