siRNA Details
Gene Symbol: Pdzk1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130401.1 | NP_569085.1 | 3, 4 | 384 | ||
XM_006238620.2 | XP_006238682.1 | 4, 5 | 405 | |||
XM_008763968.1 | XP_008762190.1 | 4, 5 | 577 | |||
XM_008763969.1 | XP_008762191.1 | 4, 5 | 405 | |||
XM_008763970.1 | XP_008762192.1 | 4, 5 | 390 | |||
GenBank mRNA | AF402772.1 | - | 3, 4 | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
