siRNA Details
Gene Symbol: Hip1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134763.1 | NP_001128235.1 | 5 | 463 | ||
NM_031234.1 | NP_112513.1 | 5 | 463 | |||
XM_008769266.1 | XP_008767488.1 | 5 | 552 | |||
XM_008769267.1 | XP_008767489.1 | 5 | 552 | |||
XM_008769268.1 | XP_008767490.1 | 5 | 2602 | |||
GenBank mRNA | AB005052.1 | - | 5 | 463 | ||
BC169096.1 | - | 5 | 450 | 7936362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
