siRNA Details
Gene Symbol: Pard3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031235.1 | NP_112514.1 | 17 | 2793 | ||
XM_006255828.1 | XP_006255890.1 | 17 | 2769 | |||
XM_006255829.1 | XP_006255891.1 | 17 | 2778 | |||
XM_006255830.1 | XP_006255892.1 | 17 | 2769 | |||
XM_006255831.1 | XP_006255893.1 | 16 | 2694 | |||
XM_006255835.1 | XP_006255897.1 | 17 | 2778 | |||
XM_006255836.1 | XP_006255898.1 | 17 | 2769 | |||
GenBank mRNA | AB005549.1 | - | 17 | 2793 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001013581, NM_033620, AK137690, AY856082, BC145508, and others.
Gene Ontology Categories:
