siRNA Details
Gene Symbol: Fut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031236.1 | NP_112515.1 | 2 | 1116 | ||
XM_008759420.1 | XP_008757642.1 | 3 | 1199 | |||
GenBank mRNA | AB006137.1 | - | 2 | 1116 | ||
L26009.1 | - | 1 | 131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
