siRNA Details
Gene Symbol: Ube2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031237.1 | NP_112516.1 | 5, 6 | 582 | ||
XM_006233333.1 | XP_006233395.1 | 6, 7 | 574 | |||
XM_006233334.1 | XP_006233396.1 | 6, 7 | 721 | |||
XM_006233335.1 | XP_006233397.1 | 6, 7 | 528 | |||
XM_006233336.2 | XP_006233398.1 | 6, 7 | 574 | |||
XM_006233337.1 | XP_006233399.1 | 6, 7 | 721 | |||
XM_006233338.1 | XP_006233400.1 | 6, 7 | 559 | |||
GenBank mRNA | AB006852.1 | - | 5, 6 | 582 | ||
AY724500.1 | - | 6, 7 | 529 | |||
BC072696.1 | - | 6, 7 | 505 | 7105617 | ||
FQ215509.1 | - | 1, 2 | 109 | |||
FQ226278.1 | - | 6, 7 | 558 | |||
FQ233355.1 | - | 6, 7 | 549 | |||
U13175.1 | - | 5, 6 | 436 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025356, AK153410, BC098220, AK009276, AK167801, and others.
Gene Ontology Categories:
