siRNA Details
Gene Symbol: Celsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191110.1 | NP_001178039.1 | 22 | 7024 | ||
XM_006233192.2 | XP_006233254.1 | 22 | 7554 | |||
GenBank mRNA | AB011529.1 | - | 22 | 4699 |
