siRNA Details
Gene Symbol: Slit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031321.1 | NP_112611.1 | 34 | 4205 | ||
XM_008767597.1 | XP_008765819.1 | 33 | 3746 | |||
GenBank mRNA | AB011531.1 | - | 34 | 4205 |
