siRNA Details
Gene Symbol: Lrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031322.3 | NP_112612.2 | 30 | 4577 | ||
XM_008762038.1 | XP_008760260.1 | 30 | 4789 | |||
GenBank mRNA | AB011533.1 | - | 17 | 2743 | ||
AB073317.3 | - | 30 | 4577 |
