siRNA Details
Gene Symbol: Lrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031322.3 | NP_112612.2 | 20, 21 | 2835 | ||
XM_008762038.1 | XP_008760260.1 | 20, 21 | 3047 | |||
GenBank mRNA | AB011533.1 | - | 7, 8 | 1001 | ||
AB073317.3 | - | 20, 21 | 2835 |
