siRNA Details
Gene Symbol: Ocln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031329.2 | NP_112619.2 | 3 | 665 | ||
XM_006231853.2 | XP_006231915.1 | 3 | 661 | |||
XM_006231854.2 | XP_006231916.1 | 3 | 705 | |||
XM_006231855.2 | XP_006231917.1 | 2 | 585 | |||
GenBank mRNA | AY033773.1 | - | 1 | 129 | ||
BC062037.1 | - | 3 | 665 | 5623744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
