siRNA Details
Gene Symbol: Hnrnpab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031330.2 | NP_112620.2 | 3 | 507 | ||
XM_003750817.3 | XP_003750865.1 | 3 | 561 | |||
GenBank mRNA | AF216753.1 | - | 2 | 308 | ||
AJ238854.1 | - | 3 | 519 | |||
AJ238855.1 | - | 3 | 519 | |||
BC066664.1 | - | 3 | 502 | 5621982 |
Gene Symbol: LOC103689931 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246345.2 | XP_006246407.1 | 3 | 561 | ||
XM_008767732.1 | XP_008765954.1 | 3 | 561 |

More Information
Other Species Targeted

Gene Ontology Categories:
