siRNA Details
Gene Symbol: Polr2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031335.3 | NP_112625.1 | 3 | 214 | ||
GenBank mRNA | AB017711.1 | - | 3 | 214 | ||
BC083552.1 | - | 3 | 254 | 7190444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
