siRNA Details
Gene Symbol: Slc7a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031341.1 | NP_112631.1 | 4 | 907 | ||
XM_006252020.2 | XP_006252082.1 | 4 | 843 | |||
XM_006252021.2 | XP_006252083.1 | 7 | 1312 | |||
XM_006252023.2 | XP_006252085.1 | 4 | 848 | |||
XM_006252024.2 | XP_006252086.1 | 4 | 1537 | |||
XM_008770705.1 | XP_008768927.1 | 6 | 1255 | |||
GenBank mRNA | AB020520.1 | - | 4 | 907 | ||
AF200684.1 | - | 3 | 715 | |||
BC091142.1 | - | 4 | 822 | 7129657 |
