siRNA Details
Gene Symbol: Ppargc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031347.1 | NP_112637.1 | 8 | 1249 | ||
XM_008770219.1 | XP_008768441.1 | 8 | 1316 | |||
XM_008770220.1 | XP_008768442.1 | 7 | 2486 | |||
GenBank mRNA | AB025784.1 | - | 8 | 1249 | ||
AY237127.1 | - | 8 | 1208 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008904, AF049330, AB061324, AK138668, BC066868, and others.
Gene Ontology Categories:
