siRNA Details
Gene Symbol: Pex3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031350.1 | NP_112640.1 | 4 | 401 | ||
GenBank mRNA | AB035306.1 | - | 4 | 401 | ||
BC062046.1 | - | 4 | 492 | 5597999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
