siRNA Details
Gene Symbol: Pex3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031350.1 | NP_112640.1 | 5 | 499 | ||
GenBank mRNA | AB035306.1 | - | 5 | 499 | ||
BC062046.1 | - | 5 | 590 | 5597999 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001164195, NM_019961, BC033415, BC037606, AK005978, and others.
Gene Ontology Categories:
