siRNA Details
Gene Symbol: Rrad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053338.2 | NP_445790.2 | 5 | 912 | ||
XM_006255055.2 | XP_006255117.1 | 5 | 994 | |||
GenBank mRNA | FM055705.1 | - | 2 | 253 | ||
U12187.1 | - | 5 | 881 |
