siRNA Details
Gene Symbol: Vdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031353.1 | NP_112643.1 | 5 | 362 | ||
XM_006246346.1 | XP_006246408.1 | 4 | 335 | |||
GenBank mRNA | AB039662.1 | - | 5 | 362 | ||
AF048828.1 | - | 5 | 352 | |||
AF268467.1 | - | 4 | 296 | |||
BC060558.1 | - | 5 | 339 | |||
BC072484.1 | - | 5 | 346 | 7105146 | ||
BC087573.1 | - | 5 | 341 | 7304939 | ||
BC087657.1 | - | 5 | 318 | 7312189 | ||
BC104684.1 | - | 5 | 332 | 7462139 | ||
BC127491.1 | - | 5 | 318 | 8370617 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
