siRNA Details
Gene Symbol: Tpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031357.1 | NP_112647.1 | 7 | 712 | ||
GenBank mRNA | AB043870.1 | - | 7 | 712 | ||
AY724484.1 | - | 7 | 713 | |||
BC081775.1 | - | 7 | 721 | 7107183 |

More Information
Other Species Targeted

Gene Ontology Categories:
