siRNA Details
Gene Symbol: Tpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031357.1 | NP_112647.1 | 3, 4 | 236 | ||
GenBank mRNA | AB043870.1 | - | 3, 4 | 236 | ||
AY724484.1 | - | 3, 4 | 237 | |||
BC081775.1 | - | 3, 4 | 245 | 7107183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
