siRNA Details
Gene Symbol: Cdkn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031762.3 | NP_113950.2 | 2 | 264 | ||
XM_006237539.2 | XP_006237601.1 | 1 | 717 | |||
GenBank mRNA | AF015194.1 | - | 1 | 188 | ||
AY623024.1 | - | 1 | 610 | |||
AY623040.1 | - | 2 | 264 | |||
D83792.1 | - | 1 | 550 | |||
D86924.1 | - | 1 | 188 |

More Information
Other Species Targeted

Gene Ontology Categories:
