siRNA Details
Gene Symbol: Rxrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031765.1 | NP_113953.1 | 4 | 757 | ||
XM_006250212.2 | XP_006250274.1 | 3 | 901 | |||
GenBank mRNA | AJ223083.1 | - | 3 | 292 | ||
BC091363.1 | - | 4 | 757 | 7319081 |
