siRNA Details
Gene Symbol: Sort1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031767.1 | NP_113955.1 | 16, 17 | 2157 | ||
XM_006233194.2 | XP_006233256.1 | 16, 17 | 2918 | |||
XM_006233195.2 | XP_006233257.1 | 16, 17 | 2916 | |||
XM_006233196.2 | XP_006233258.1 | 16, 17 | 2912 | |||
GenBank mRNA | AF019109.1 | - | 16, 17 | 1870 | ||
AF023621.1 | - | 14, 15 | 1771 |
