siRNA Details
Gene Symbol: Itgae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031768.2 | NP_113956.2 | 26 | 3014 | ||
GenBank mRNA | AF020046.1 | - | 26 | 2983 |

More Information
Other Species Targeted

Gene Ontology Categories:
