siRNA Details
Gene Symbol: Gda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031776.2 | NP_113964.2 | 12 | 1197 | ||
GenBank mRNA | AF026472.2 | - | 12 | 1254 | ||
AF245172.1 | - | 12 | 1197 | |||
FQ233257.1 | - | 12 | 1220 | |||
FQ233523.1 | - | 12 | 1225 |
