siRNA Details
Gene Symbol: Slc32a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031782.1 | NP_113970.1 | 1 | 234 | ||
XM_006235441.2 | XP_006235503.1 | 1 | 398 | |||
GenBank mRNA | AF030253.1 | - | 1 | 234 |

More Information
Other Species Targeted

Gene Ontology Categories:
