siRNA Details
Gene Symbol: Nefl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031783.1 | NP_113971.1 | 2 | 1230 | ||
GenBank mRNA | AF031880.1 | - | 2 | 1230 | ||
M25638.1 | - | 2 | 568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
