siRNA Details
Gene Symbol: STYX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130701.1 | NP_001124173.1 | 6 | 325 | ||
NM_145251.3 | NP_660294.1 | 5 | 573 | |||
XM_006720242.3 | XP_006720305.1 | 3 | 193 | |||
XM_011537108.2 | XP_011535410.1 | 5 | 240 | |||
GenBank mRNA | AK098195.1 | - | 5 | 556 | ||
AL832591.1 | - | 6 | 310 | |||
BC020265.2 | - | 5 | 524 | 3922829 | ||
BC146995.1 | - | 5 | 463 | 9056923 | ||
BC146998.1 | - | 5 | 463 | 9056926 | ||
BX647636.1 | - | 5 | 531 |
