siRNA Details
Gene Symbol: Pias3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031784.2 | NP_113972.2 | 9, 10 | 1196 | ||
XM_006232991.2 | XP_006233053.1 | 9, 10 | 1449 | |||
XM_006232992.2 | XP_006233054.1 | 9, 10 | 2531 | |||
XM_006232993.2 | XP_006233055.1 | 9, 10 | 1338 | |||
XM_008761337.1 | XP_008759559.1 | 9, 10 | 1200 | |||
XM_008761338.1 | XP_008759560.1 | 9, 10 | 1449 | |||
GenBank mRNA | AF032872.2 | - | 9, 10 | 1196 |
