siRNA Details
Gene Symbol: Pias3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031784.2 | NP_113972.2 | 9 | 1120 | ||
XM_006232991.2 | XP_006233053.1 | 9 | 1373 | |||
XM_006232992.2 | XP_006233054.1 | 9 | 2455 | |||
XM_006232993.2 | XP_006233055.1 | 9 | 1262 | |||
XM_008761337.1 | XP_008759559.1 | 9 | 1124 | |||
XM_008761338.1 | XP_008759560.1 | 9 | 1373 | |||
GenBank mRNA | AF032872.2 | - | 9 | 1120 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001165949, NM_146135, NM_018812, BC051252, AK134404, and others.
Gene Ontology Categories:
