siRNA Details
Gene Symbol: Spag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031792.1 | NP_113980.1 | 11 | 1223 | ||
XM_008762386.1 | XP_008760608.1 | 11 | 957 | |||
GenBank mRNA | AF043345.2 | - | 11 | 1223 |
Gene Symbol: LOC100911109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749611.3 | XP_003749659.1 | 1365 | |||
XM_008762411.1 | XP_008760633.1 | 957 |
