siRNA Details
Gene Symbol: Gpr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031806.1 | NP_113994.1 | 2 | 633 | ||
GenBank mRNA | AF064706.1 | - | 2 | 633 | ||
U12006.1 | - | 2 | 633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
