siRNA Details
Gene Symbol: Cngb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031809.1 | NP_113997.1 | 26 | 2937 | ||
XM_006255124.2 | XP_006255186.1 | 25 | 2629 | |||
GenBank mRNA | AF015728.1 | - | 3 | 272 | ||
AF068572.1 | - | 14 | 1969 | |||
AJ000496.1 | - | 26 | 2937 | |||
AJ000515.1 | - | 14 | 1871 |
